Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ripply2
Entrez Gene ID
100761028
Gene Name
ripply transcriptional repressor 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613597: 3716481 - 3720688 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495634.1 XP_003495682.1 3 - 4 233 121

View Full Product Details