Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cactin
Entrez Gene ID
100771890
Gene Name
cactin, spliceosome C complex subunit
Gene Aliases
I79_013837
Chromosome Location
Chr.Un|NW_003614244: 540923 - 547206 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646986.1 XP_007645176.1 7 - 8 1446 98

View Full Product Details