Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olig3
Entrez Gene ID
100772209
Gene Name
oligodendrocyte transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613627: 1936914 - 1937735 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496546.1 XP_003496594.1 1 - 1 620 98

View Full Product Details