Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpeb2
Entrez Gene ID
100761763
Gene Name
cytoplasmic polyadenylation element binding protein 2
Gene Aliases
I79_014261
Chromosome Location
Chr.Un|NW_003614289: 281760 - 331408 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506782.2 XP_003506830.1 8 - 9 936 89

View Full Product Details