Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bmp3
Entrez Gene ID
100766849
Gene Name
bone morphogenetic protein 3
Gene Aliases
I79_023784
Chromosome Location
Chr.Un|NW_003616697: 28417 - 60348 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514536.1 XP_003514584.1 3 - 4 553 67

View Full Product Details