Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slamf9
Entrez Gene ID
100755075
Gene Name
SLAM family member 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613783: 963625 - 966366 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640975.1 XP_007639165.1 3 - 4 726 80

View Full Product Details