Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Znf45
Entrez Gene ID
100767782
Gene Name
zinc finger protein 45
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614529: 123181 - 140221 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648996.1 XP_007647186.1 7 - 8 2011 89

View Full Product Details