Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vopp1
Entrez Gene ID
100767477
Gene Name
vesicular, overexpressed in cancer, prosurvival protein 1
Gene Aliases
I79_012031
Chromosome Location
Chr.Un|NW_003614101: 896042 - 952984 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645395.1 XP_007643585.1 1 - 2 196 97

View Full Product Details