Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lmo1
Entrez Gene ID
100768173
Gene Name
LIM domain only 1 (rhombotin 1)
Gene Aliases
I79_018946
Chromosome Location
Chr.Un|NW_003614918: 182377 - 196064 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651229.1 XP_007649419.1 3 - 4 362 89

View Full Product Details