Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scgb3a2
Entrez Gene ID
100754330
Gene Name
secretoglobin, family 3A, member 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614509: 243204 - 246205 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508256.1 XP_003508304.1 1 - 2 54 76

View Full Product Details