Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cgref1
Entrez Gene ID
100771935
Gene Name
cell growth regulator with EF-hand domain 1
Gene Aliases
I79_002567
Chromosome Location
Chr.Un|NW_003613640: 2130618 - 2133388 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007655209.1 XP_007653399.1 2 - 3 151 77

View Full Product Details