Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grpr
Entrez Gene ID
100758098
Gene Name
gastrin-releasing peptide receptor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614454: 188556 - 222081 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507926.2 XP_003507974.1 1 - 2 786 76

View Full Product Details