Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab38
Entrez Gene ID
100768866
Gene Name
RAB38, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615024: 16761 - 79959 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511080.1 XP_003511128.1 1 - 2 204 80

View Full Product Details