Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tysnd1
Entrez Gene ID
100769469
Gene Name
trypsin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613759: 1075177 - 1083202 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640646.1 XP_007638836.1 3 - 4 634 64

View Full Product Details