Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ss18l2
Entrez Gene ID
100771870
Gene Name
synovial sarcoma translocation gene on chromosome 18-like 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613803: 1169124 - 1171766 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500690.2 XP_003500738.1 2 - 3 245 109

View Full Product Details