Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam71e2
Entrez Gene ID
100773246
Gene Name
family with sequence similarity 71, member E2
Gene Aliases
I79_016108
Chromosome Location
Chr.Un|NW_003614505: 138253 - 151962 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648863.1 XP_007647053.1 4 - 5 283 86

View Full Product Details