Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Inhbb
Entrez Gene ID
100759312
Gene Name
inhibin, beta B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613737: 2343676 - 2350931 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499438.2 XP_003499486.1 1 - 2 77 73

View Full Product Details