Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sap25
Entrez Gene ID
100755701
Gene Name
Sin3A-associated protein, 25kDa
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614921: 177897 - 179504 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651253.1 XP_007649443.1 5 - 6 740 66

View Full Product Details