Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phyhip
Entrez Gene ID
100767230
Gene Name
phytanoyl-CoA 2-hydroxylase interacting protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613613: 1865409 - 1870887 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496164.1 XP_003496212.1 1 - 2 167 99

View Full Product Details