Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nxph1
Entrez Gene ID
100759051
Gene Name
neurexophilin 1
Gene Aliases
I79_010818
Chromosome Location
Chr.Un|NW_003614010: 69457 - 81853 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644440.1 XP_007642630.1 1 - 2 154 86

View Full Product Details