Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nkx2-3
Entrez Gene ID
100768689
Gene Name
NK2 homeobox 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613664: 2998696 - 3001074 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497789.1 XP_003497837.1 2 - 3 539 104

View Full Product Details