Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Caps
Entrez Gene ID
100759707
Gene Name
calcyphosine
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613861: 907607 - 910490 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501758.2 XP_003501806.2 2 - 3 1285 64

View Full Product Details