Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bcl2l2
Entrez Gene ID
103159537
Gene Name
BCL2-like 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614213: 948942 - 953631 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646593.1 XP_007644783.1 2 - 3 638 62

View Full Product Details