Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tax1bp3
Entrez Gene ID
100772000
Gene Name
Tax1 (human T-cell leukemia virus type I) binding protein 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614665: 410151 - 412201 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649776.1 XP_007647966.1 1 - 2 213 78
GenBank mRNA JP050900.1 - 1 - 2 213 78

View Full Product Details