Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myeov2
Entrez Gene ID
100769752
Gene Name
myeloma overexpressed 2
Gene Aliases
I79_005294
Chromosome Location
Chr.Un|NW_003613726: 242237 - 246243 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499251.2 XP_003499299.1 2 - 3 174 95
GenBank mRNA JI882748.1 - 2 - 3 172 95

View Full Product Details