Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfhx2
Entrez Gene ID
100764649
Gene Name
zinc finger homeobox 2
Gene Aliases
I79_008565
Chromosome Location
Chr.Un|NW_003613875: 1646212 - 1678772 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502073.2 XP_003502121.1 3 - 4 2929 98

View Full Product Details