Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cenpv
Entrez Gene ID
100761123
Gene Name
centromere protein V
Gene Aliases
I79_001748
Chromosome Location
Chr.Un|NW_003613619: 2145919 - 2179248 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649487.1 XP_007647677.1 3 - 4 462 83
GenBank mRNA JI882997.1 - 1 - 2 309 83

View Full Product Details