Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sptb
Entrez Gene ID
100765992
Gene Name
spectrin, beta, erythrocytic
Gene Aliases
I79_005365, Spnb1
Chromosome Location
Chr.Un|NW_003613728: 2383799 - 2453971 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499317.1 XP_003499365.1 20 - 21 4469 56

View Full Product Details