Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Znrf2
Entrez Gene ID
100760215
Gene Name
zinc and ring finger 2
Gene Aliases
I79_015529
Chromosome Location
Chr.Un|NW_003614428: 676166 - 704324 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507796.2 XP_003507844.1 1 - 2 40 103

View Full Product Details