Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Esm1
Entrez Gene ID
100773837
Gene Name
endothelial cell-specific molecule 1
Gene Aliases
I79_020754
Chromosome Location
Chr.Un|NW_003615347: 85963 - 93256 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652559.1 XP_007650749.1 1 - 2 303 70

View Full Product Details