Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lin28b
Entrez Gene ID
100770182
Gene Name
lin-28 homolog B (C. elegans)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614750: 84085 - 165247 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509668.1 XP_003509716.1 1 - 2 106 76

View Full Product Details