Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Serinc4
Entrez Gene ID
100766569
Gene Name
serine incorporator 4
Gene Aliases
I79_003759
Chromosome Location
Chr.Un|NW_003613677: 848125 - 853786 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007622386.1 XP_007620576.1 7 - 8 985 79

View Full Product Details