Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ak6
Entrez Gene ID
100768838
Gene Name
adenylate kinase 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614223: 241690 - 257498 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506264.2 XP_003506312.1 4 - 5 383 121
GenBank mRNA JI869439.1 - 2 - 3 196 121

View Full Product Details