Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zmat1
Entrez Gene ID
100754864
Gene Name
zinc finger, matrin-type 1
Gene Aliases
I79_023891
Chromosome Location
Chr.Un|NW_003616787: 31863 - 89789 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654703.1 XP_007652893.1 7 - 8 904 80
XM_007654704.1 XP_007652894.1 7 - 8 895 80

View Full Product Details