Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aqp10
Entrez Gene ID
100751558
Gene Name
aquaporin 10
Gene Aliases
I79_008034
Chromosome Location
Chr.Un|NW_003613854: 1133805 - 1137400 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501658.1 XP_003501706.1 5 - 6 704 97

View Full Product Details