Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hapln4
Entrez Gene ID
100764042
Gene Name
hyaluronan and proteoglycan link protein 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613716: 389732 - 400159 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007638344.1 XP_007636534.1 4 - 5 649 88

View Full Product Details