Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nms
Entrez Gene ID
100757218
Gene Name
neuromedin S
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613959: 564994 - 576025 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503334.1 XP_003503382.1 1 - 2 81 76

View Full Product Details