Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cmtm4
Entrez Gene ID
100768129
Gene Name
CKLF-like MARVEL transmembrane domain containing 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613750: 177296 - 195266 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499685.2 XP_003499733.1 1 - 2 84 77

View Full Product Details