Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zadh2
Entrez Gene ID
100762407
Gene Name
zinc binding alcohol dehydrogenase domain containing 2
Gene Aliases
I79_024680
Chromosome Location
Chr.Un|NW_003617554: 38528 - 47761 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007655211.1 XP_007653401.1 1 - 2 48 75

View Full Product Details