Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tex22
Entrez Gene ID
100754201
Gene Name
testis expressed 22
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613702: 2659533 - 2662559 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498772.2 XP_003498820.1 1 - 2 30 83

View Full Product Details