Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lacc1
Entrez Gene ID
100769755
Gene Name
laccase (multicopper oxidoreductase) domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613785: 1339686 - 1368982 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500302.2 XP_003500350.1 4 - 5 1169 73
XM_007641011.1 XP_007639201.1 4 - 5 1169 73
XM_007641012.1 XP_007639202.1 4 - 5 1169 73
XM_007641013.1 XP_007639203.1 4 - 5 1169 73
GenBank mRNA JI863040.1 - 4 - 5 1126 73
JP057077.1 - 4 - 5 1126 73

View Full Product Details