Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Samd7
Entrez Gene ID
100767182
Gene Name
sterile alpha motif domain containing 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613976: 553335 - 565565 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503499.1 XP_003503547.1 4 - 5 894 60

View Full Product Details