Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxf1
Entrez Gene ID
100757169
Gene Name
forkhead box F1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613583: 2461380 - 2466786 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007609980.1 XP_007608170.1 1 - 2 23 74

View Full Product Details