Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mogat1
Entrez Gene ID
100761111
Gene Name
monoacylglycerol O-acyltransferase 1
Gene Aliases
I79_021091
Chromosome Location
Chr.Un|NW_003615425: 142908 - 170842 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652838.1 XP_007651028.1 1 - 2 1057 79

View Full Product Details