Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc69
Entrez Gene ID
100767187
Gene Name
leucine rich repeat containing 69
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614081: 121686 - 234668 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504686.2 XP_003504734.1 3 - 4 383 120

View Full Product Details