Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ntsr1
Entrez Gene ID
100756437
Gene Name
neurotensin receptor 1 (high affinity)
Gene Aliases
I79_009865
Chromosome Location
Chr.Un|NW_003613943: 430912 - 471759 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503117.1 XP_003503165.1 1 - 2 712 89

View Full Product Details