Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx39a
Entrez Gene ID
100752425
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A
Gene Aliases
Ddx39, I79_008391
Chromosome Location
Chr.Un|NW_003613865: 1556692 - 1566133 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501883.2 XP_003501931.1 8 - 9 1071 74
GenBank mRNA JI892008.1 - 2 - 3 237 74
JP055369.1 - 3 - 4 298 74

View Full Product Details