Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnk1
Entrez Gene ID
100762483
Gene Name
potassium channel, subfamily K, member 1
Gene Aliases
I79_002279
Chromosome Location
Chr.Un|NW_003613636: 990635 - 1024353 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496802.1 XP_003496850.1 1 - 2 354 83

View Full Product Details