Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Golt1a
Entrez Gene ID
100768539
Gene Name
golgi transport 1A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614184: 284607 - 295221 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505764.1 XP_003505812.1 2 - 3 119 72

View Full Product Details