Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pfdn6
Entrez Gene ID
100750607
Gene Name
prefoldin subunit 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614972: 366570 - 367870 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510832.2 XP_003510880.1 1 - 2 172 71
GenBank mRNA JI893460.1 - 1 - 2 134 71
JP038674.1 - 1 - 2 134 71

View Full Product Details